Learn More
Selleck Chemical LLC Avobenzone S1904-25mg
SDP
Catalog No. 503110798
Encompass_Preferred
Avobenzone (Butyl methoxydibenzoylmethane BF2AVB) is an oil soluble ingredient used in sunscreen products to absorb the full spectrum of UVA rays and a dibenzoylmethane derivative
Catalog No. 50-311-0798 Supplier Selleck Chemical LLC Supplier No. S190425MG
Avobenzone (Butyl methoxydibenzoylmethane BF2AVB) is an oil soluble ingredient used in sunscreen products to absorb the full spectrum of UVA rays and a dibenzoylmethane derivative