Learn More
U.S. Pharmacopeia Avobenzone, 70356-09-1, MFCD00210252, 500mg
Catalog No. NC9097129
Encompass
Molecular Formula C20H22O3, Molecular Weight 310.4, Melting Point 177.8 - 186.8 °F (81 - 86 °C), Flash Point > 200.00 °F (> 93.33 °C) Tag Closed Cup, Synonyms: 1-[4-(1,1-dimethylethyl)phenyl]-3-(4-methoxyphenyl)-1,3-Propanedione, Butyl methoxydibenzoylmethane
Catalog No. NC9097129 Supplier U.S. Pharmacopeia Supplier No. 1045337
Molecular Formula C20H22O3, Molecular Weight 310.4, Melting Point 177.8 - 186.8 °F (81 - 86 °C), Flash Point > 200.00 °F (> 93.33 °C) Tag Closed Cup, Synonyms: 1-[4-(1,1-dimethylethyl)phenyl]-3-(4-methoxyphenyl)-1,3-Propanedione, Butyl methoxydibenzoylmethane