Learn More
Apexbio Technology LLC Avobenzone 70356-09-1 200mg
SDP
Catalog No. 5000091067
Encompass_Preferred
Avobenzone (70356-09-1) is a small-molecule agonist targeting estrogen receptor (ER ) It is designed to bind directly to ER thereby modulating endocrine signaling pathways Avobenzone exerts its biological activity primarily through direct receptor binding and estrogenic agonist activity Based on these properties Avobenzone holds research potential in studies investigating endocrine-disrupting chemicals mechanisms of photoprotection UV-induced cellular damage and skin pathology
Catalog No. 50-000-91067 Supplier Apexbio Technology LLC Supplier No. B2079200
Avobenzone (70356-09-1) is a small-molecule agonist targeting estrogen receptor (ER ) It is designed to bind directly to ER thereby modulating endocrine signaling pathways Avobenzone exerts its biological activity primarily through direct receptor binding and estrogenic agonist activity Based on these properties Avobenzone holds research potential in studies investigating endocrine-disrupting chemicals mechanisms of photoprotection UV-induced cellular damage and skin pathology