Learn More
Medchemexpress LLC Avobenzone | 70356-09-1 | 98.0% | 10 G
SDP
Catalog No. 5000406902
Encompass_Preferred
Avobenzone is a dibenzoylmethane compound widely used as a filter in sunscreens for skin photoprotection in the UVA band. It is also identified as an endocrine disruptor that directly binds to estrogen receptor β and acts as an estrogen agonist.
  • Widely used as a filter in sunscreens for UVA band skin photoprotection
  • Acts as an endocrine disruptor and estrogen agonist
  • Promotes adipogenesis in hBM-MSCs
  • Up-regulates mRNA levels of PPARγ during adipogenesis
  • Inhibits proliferative activities of human trophoblast cells
  • Induces apoptosis in HTR8/SVneo cells
  • Exhibits weak ERα agonism and weak AR antagonism
Catalog No. 50-004-06902 Supplier Medchemexpress LLC Supplier No. HYB031610G
Avobenzone is a dibenzoylmethane compound widely used as a filter in sunscreens for skin photoprotection in the UVA band. It is also identified as an endocrine disruptor that directly binds to estrogen receptor β and acts as an estrogen agonist.
  • Widely used as a filter in sunscreens for UVA band skin photoprotection
  • Acts as an endocrine disruptor and estrogen agonist
  • Promotes adipogenesis in hBM-MSCs
  • Up-regulates mRNA levels of PPARγ during adipogenesis
  • Inhibits proliferative activities of human trophoblast cells
  • Induces apoptosis in HTR8/SVneo cells
  • Exhibits weak ERα agonism and weak AR antagonism