Learn More
Medchemexpress LLC Avobenzone | 70356-09-1 | 98.0% | 5 G
SDP
Catalog No. 5000406660
Encompass_Preferred
Avobenzone, a dibenzoylmethane compound, is one of the most widely used filters in sunscreens for skin photoprotection in the UVA band. It is also an endocrine disruptor that directly binds to estrogen receptor β and acts as an estrogen agonist.
  • Promotes adipogenesis in hBM-MSCs
  • Up-regulates mRNA levels of PPARγ during adipogenesis
  • Inhibits proliferative activities of human trophoblast cells
  • Induces apoptosis in HTR8/SVneo cells
  • Shows weak ERα agonism and weak AR antagonism
Catalog No. 50-004-06660 Supplier Medchemexpress LLC Supplier No. HYB03165G
Avobenzone, a dibenzoylmethane compound, is one of the most widely used filters in sunscreens for skin photoprotection in the UVA band. It is also an endocrine disruptor that directly binds to estrogen receptor β and acts as an estrogen agonist.
  • Promotes adipogenesis in hBM-MSCs
  • Up-regulates mRNA levels of PPARγ during adipogenesis
  • Inhibits proliferative activities of human trophoblast cells
  • Induces apoptosis in HTR8/SVneo cells
  • Shows weak ERα agonism and weak AR antagonism