Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avobenzone, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018526 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8526 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8526 Supplier MilliporeSigma Supelco Supplier No. 1663325MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Avobenzone is an UVA filter. It is used in sunscreen lotions and cosmetic formulations. It has maximum absorption at ca 340-350 nm, decreasing under UV irradiation resulting in loss of the UVA protecting effect. Its photostability is very sensitive as it is stable in polar protic solvent and is photolabile in nonpolar solvents. Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 70356-09-1
Physical Form Neat
Linear Formula C20H22O3
MDL Number MFCD00210252
Melting Point 81°C to 84°C
Synonym 1-(4-Methoxyphenyl)-3-(4-tert-butylphenyl)-1,3-propanedione; Butyl methoxydibenzoylmethane
Type Analytical Standard
Quantity 25 mg
Formula Weight 310.39
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.