Learn More
Medchemexpress LLC Avotaciclib hydrochloride | 00-00-0 | 99.8% | 317.73 | C13H12ClN7O | 5 MG
SDP
Catalog No. 5000221798
Encompass_Preferred
Avotaciclib hydrochloride is the hydrochloride salt of avotaciclib, an orally active cyclin-dependent kinase 1 (CDK1) inhibitor used in preclinical cancer research. It inhibits tumor cell proliferation and can induce apoptosis, and is supplied as a solid for laboratory studies.
  • Potent CDK1 inhibitor suitable for in vitro and preclinical studies.
  • Hydrochloride salt form for improved solubility.
  • Chemical formula C13H12ClN7O, molecular weight 317.73.
  • High purity (approximately 99.8%) suitable for research applications.
  • Supplied as a 5 mg solid pack.
  • Used in research on pancreatic and lung cancer models.
Catalog No. 50-002-21798 Supplier Medchemexpress LLC Supplier No. HY137432B5MG
Avotaciclib hydrochloride is the hydrochloride salt of avotaciclib, an orally active cyclin-dependent kinase 1 (CDK1) inhibitor used in preclinical cancer research. It inhibits tumor cell proliferation and can induce apoptosis, and is supplied as a solid for laboratory studies.
  • Potent CDK1 inhibitor suitable for in vitro and preclinical studies.
  • Hydrochloride salt form for improved solubility.
  • Chemical formula C13H12ClN7O, molecular weight 317.73.
  • High purity (approximately 99.8%) suitable for research applications.
  • Supplied as a 5 mg solid pack.
  • Used in research on pancreatic and lung cancer models.