Learn More
Medchemexpress LLC Avotaciclib hydrochloride | 99.8% | 317.73 | 25 MG
Supplier: Medchemexpress LLC 25MGHY137432B
Avotaciclib hydrochloride is an orally active cyclin-dependent kinase 1 (CDK1) inhibitor. It inhibits the proliferation and induces apoptosis of tumor cells. This compound is used in the research of various cancers, including pancreatic cancer and lung cancer.
- Orally active cyclin-dependent kinase 1 (CDK1) inhibitor
- Inhibits proliferation of tumor cells
- Induces apoptosis of tumor cells
- Useful in the research of pancreatic cancer and lung cancer
- Inhibits cell proliferation and promotes apoptosis in radiotherapy-resistant non-small cell lung cancer cell lines
- Demonstrated enhanced effects when used in combination therapies