Learn More
Aviva Systems Biology Corp AVPR1A Antibody - C-terminal region (ARP62992_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62992_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AVPR1A. It was validated on Western Blot. . Alternate names: AVPR1ApplicationWB. Species reactivity: Guinea Pig, Human, Pig, Rabbit, Rat
Catalog No. 50-287-833