Learn More
New England Biolabs, Inc. AvrII – 100 units
SDP
Catalog No. 50812087
Encompass
A restriction endonuclease that recognizes the sequence C^CTAG_G.
Catalog No. 50-812-087 Supplier New England Biolabs, Inc. Supplier No. R0174S
A restriction endonuclease that recognizes the sequence C^CTAG_G.