Learn More
Cayman Chemical Avermectin B1a monosaccharide, Purity: ≥85%, CAS: 71831-09-9, 5 mg
SDP
Catalog No. 502934177
Encompass_Preferred
A macrolide anthelmintic; lethal to C. elegans (MAC = 0.1 μM); stimulates conductance in isolated shore crab muscle
Catalog No. 50-293-4177 Supplier Cayman Chemical Supplier No. 266905 MG
A macrolide anthelmintic; lethal to C. elegans (MAC = 0.1 μM); stimulates conductance in isolated shore crab muscle