Learn More
Julabo USA Inc Each, AWC100 circulator
SDP
Catalog No. 507533987
Encompass_Preferred
Chillers, Julabo, AWC100 Compact Recirculating Cooler, The AWC100, Air to Water Recirculating Cooler is a compact solution designed to remove small heat loads from external systems such as from Peltier-elements. The AWC100 features a compact footprint and is designed specifically for applications where cooling water can be above ambient and where temperature control is secondary. The ventilator speed can be increased by changing the switch for booster cooling on the rear of the unit. Stage 0: normal cooling capacity (to 400W), virtually noiseless. Stage 1: higher cooling capacity, higher ventilator speed (to 550W).Included with each unit: 2 barbed fittings for tubing 8 and 10 mm inner dia. (pump connections M10x1female)
Catalog No. 507533987 Supplier Julabo USA Inc Supplier No. 9630100.99
Chillers, Julabo, AWC100 Compact Recirculating Cooler, The AWC100, Air to Water Recirculating Cooler is a compact solution designed to remove small heat loads from external systems such as from Peltier-elements. The AWC100 features a compact footprint and is designed specifically for applications where cooling water can be above ambient and where temperature control is secondary. The ventilator speed can be increased by changing the switch for booster cooling on the rear of the unit. Stage 0: normal cooling capacity (to 400W), virtually noiseless. Stage 1: higher cooling capacity, higher ventilator speed (to 550W).Included with each unit: 2 barbed fittings for tubing 8 and 10 mm inner dia. (pump connections M10x1female)