Learn More
Cayman Chemical AWD 131-138, Purity: ≥98%, CAS: 188116-07-6, 50 mg
SDP
Catalog No. 502970115
Encompass_Preferred
A partial agonist of the GABAA receptor that acts at the benzodiazepine binding site, dose-dependently stimulating GABA currents with a threshold of 0.3-1.0 µM; displays a broad spectrum of anticonvulsant activity in diverse seizure and epilepsy models without a tendency toward tolerance or abuse
Catalog No. 50-297-0115 Supplier Cayman Chemical Supplier No. 1623550 MG
A partial agonist of the GABAA receptor that acts at the benzodiazepine binding site, dose-dependently stimulating GABA currents with a threshold of 0.3-1.0 µM; displays a broad spectrum of anticonvulsant activity in diverse seizure and epilepsy models without a tendency toward tolerance or abuse