Learn More
Medchemexpress LLC HY-114415 10mg Medchemexpress, AWZ1066S CAS:2239272-16-1 Purity:>98%
SDP
Catalog No. 501969353
Encompass_Preferred
Medchemexpress, HY-114415 10mg AWZ1066S CAS:2239272-16-1 AWZ1066S is a highly specific anti-Wolbachia drug candidate for a short-course treatment of filariasis, with an EC50 of 2.5 nM in cell assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9353 Supplier Medchemexpress LLC Supplier No. HY11441510MG
Medchemexpress, HY-114415 10mg AWZ1066S CAS:2239272-16-1 AWZ1066S is a highly specific anti-Wolbachia drug candidate for a short-course treatment of filariasis, with an EC50 of 2.5 nM in cell assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.