Learn More
Medchemexpress LLC AX-024 hydrochloride | 1704801-24-0 | 99.2% | 375.86 g/mol | C21H23ClFNO2 | 1 ML
SDP
Catalog No. 5000240880
Encompass_Preferred
AX-024 hydrochloride is a small-molecule inhibitor of the T cell receptor-Nck interaction that selectively inhibits TCR-triggered T cell activation. Supplied for research use in biochemical and cell-based assays, it is available as a 10 mM solution in DMSO (1 mL) or as solids for custom preparation.
  • Selective inhibitor of TCR-Nck interaction for T cell signaling studies.
  • High purity suitable for biochemical and cell-based assays.
  • Provided as a 10 mM DMSO solution for ready-to-use experiments.
  • Also available in multiple solid sizes for dosing flexibility.
  • Store sealed at cold temperatures to preserve stability.
Catalog No. 50-002-40880 Supplier Medchemexpress LLC Supplier No. HY107390A10MM 1ML
AX-024 hydrochloride is a small-molecule inhibitor of the T cell receptor-Nck interaction that selectively inhibits TCR-triggered T cell activation. Supplied for research use in biochemical and cell-based assays, it is available as a 10 mM solution in DMSO (1 mL) or as solids for custom preparation.
  • Selective inhibitor of TCR-Nck interaction for T cell signaling studies.
  • High purity suitable for biochemical and cell-based assays.
  • Provided as a 10 mM DMSO solution for ready-to-use experiments.
  • Also available in multiple solid sizes for dosing flexibility.
  • Store sealed at cold temperatures to preserve stability.