Learn More
Cayman Chemical AX-024 (hydrochloride), Purity: ≥98%, CAS: 1704801-24-0, 10 mg
SDP
Catalog No. 502941019
Encompass_Preferred
CD3ε-Nck interaction; inhibits T cell receptor-triggered T cell activation (IC50 = 1 nM); prevents CD4+ T cell and macrophage infiltration to the cerebellum and spinal cord and improves neurological impairments in a MOG35-55 mouse model of EAE at 10 mg/kg per day; prevents the infiltration of eosinophils, macrophages, and neutrophils into the airway in an ovalbumin-sensitized mouse model of allergic airway disease at 50 mg/kg
Catalog No. 50-294-1019 Supplier Cayman Chemical Supplier No. 2733310 MG
CD3ε-Nck interaction; inhibits T cell receptor-triggered T cell activation (IC50 = 1 nM); prevents CD4+ T cell and macrophage infiltration to the cerebellum and spinal cord and improves neurological impairments in a MOG35-55 mouse model of EAE at 10 mg/kg per day; prevents the infiltration of eosinophils, macrophages, and neutrophils into the airway in an ovalbumin-sensitized mouse model of allergic airway disease at 50 mg/kg