Learn More
Medchemexpress LLC Axc-715 hydrochloride | 2490497-93-1 | 99.5% | 347.89 g/mol | C18H26ClN5 | 5 MG
SDP
Catalog No. 5000222940
Encompass_Preferred
AXC-715 hydrochloride is the hydrochloride salt of a small-molecule TLR7/TLR8 dual agonist used in preclinical research as an immunostimulant and as a building block for antibody-adjuvant immunoconjugates. It is supplied as a high-purity solid suitable for in vitro and in vivo studies, with recommended storage conditions to maintain stability.
Catalog No. 50-002-22940 Supplier Medchemexpress LLC Supplier No. HY138139A5MG
AXC-715 hydrochloride is the hydrochloride salt of a small-molecule TLR7/TLR8 dual agonist used in preclinical research as an immunostimulant and as a building block for antibody-adjuvant immunoconjugates. It is supplied as a high-purity solid suitable for in vitro and in vivo studies, with recommended storage conditions to maintain stability.