Learn More
Medchemexpress LLC Axc-715 hydrochloride | 2490497-93-1 | 99.5% | C18H26ClN5 | 10MG
SDP
Catalog No. 5000260557
Encompass_Preferred
AXC-715 hydrochloride is the hydrochloride salt of a small-molecule TLR7/TLR8 dual agonist used for preclinical immunology and oncology research. Supplied for research use only, it is described as an immune adjuvant and has been used in synthesis of antibody-adjuvant conjugates and in vivo antitumor models.
  • Dual TLR7/TLR8 agonist activity for immune-modulating studies
  • High purity (99.5%) for reproducible experimental results
  • Hydrochloride salt form improves aqueous solubility relative to the free base
  • Suitable for conjugation and formulation in antibody-adjuvant constructs
  • Available in small-scale research quantities for preclinical work
  • Storage recommendations provided for solid and solution forms to maintain stability
Catalog No. 50-002-60557 Supplier Medchemexpress LLC Supplier No. HY138139A10MG
AXC-715 hydrochloride is the hydrochloride salt of a small-molecule TLR7/TLR8 dual agonist used for preclinical immunology and oncology research. Supplied for research use only, it is described as an immune adjuvant and has been used in synthesis of antibody-adjuvant conjugates and in vivo antitumor models.
  • Dual TLR7/TLR8 agonist activity for immune-modulating studies
  • High purity (99.5%) for reproducible experimental results
  • Hydrochloride salt form improves aqueous solubility relative to the free base
  • Suitable for conjugation and formulation in antibody-adjuvant constructs
  • Available in small-scale research quantities for preclinical work
  • Storage recommendations provided for solid and solution forms to maintain stability