Learn More
Medchemexpress LLC Axelopran sulfate | 949904-50-1 | 99.1% | 555.68 | 1 MG
SDP
Catalog No. 5000393638
Encompass_Preferred
Axelopran sulfate is an opioid receptor antagonist with pKi values of 9.8, 8.8, and 9.9 for human recombinant μ and δ receptors and guinea pig κ receptor, respectively. It is provided as a solid, white to off-white compound and is soluble in DMSO.
  • Opioid receptor antagonist
  • Specific pKi values for human recombinant μ and δ receptors and guinea pig κ receptor
  • Solid appearance
  • White to off-white color
  • Soluble in DMSO: ≥ 100 mg/mL
  • For research use only
Catalog No. 50-003-93638 Supplier Medchemexpress LLC Supplier No. HY16765A1MG
Axelopran sulfate is an opioid receptor antagonist with pKi values of 9.8, 8.8, and 9.9 for human recombinant μ and δ receptors and guinea pig κ receptor, respectively. It is provided as a solid, white to off-white compound and is soluble in DMSO.
  • Opioid receptor antagonist
  • Specific pKi values for human recombinant μ and δ receptors and guinea pig κ receptor
  • Solid appearance
  • White to off-white color
  • Soluble in DMSO: ≥ 100 mg/mL
  • For research use only