Learn More
Perkin Elmer US LLC Axial Purge Extension Window for Optima 3x00/4x00/5x00/7x00 DV/8300
Catalog No. 509043808
Encompass_Preferred
The Axial View Purge Extension Window is a replaceable quartz window that slides into the purge extension between the torch and the optics.
Catalog No. 50-904-3808 Supplier Perkin Elmer US LLC Supplier No. N0771116
The Axial View Purge Extension Window is a replaceable quartz window that slides into the purge extension between the torch and the optics.