Learn More
Mitegen Axiocam 212 Microscope Camera, For Use With MiTeGen Stereo Microscope Kits
SDP
Catalog No. 5000495022
Encompass
The ZEISS Axiocam 212 is a smart 12-megapixel color microscope camera capable of delivering images at full 4K resolution and up to 30 frames per second. This camera can be used these MiTeGen Stereo Microscope Kits, Stemi 305, Stemi 508, V81, V121, or V201, when combined with the Axiocam Camera Adapter. In stand-alone mode, the camera automatically adjusts brightness and white balance and offers live image enhancement functions like sharpening, denoising, and HDR without the need for a PC. Alternatively, the CMOS camera can be connected to a computer via USB, Ethernet, or wirelessly via Wi-Fi dongle, making it an ideal solution for digital classroom applications and connected laboratories. SKU for adapter is ZV-Erc5-Adapter, and is necessary to mount the Axiocam 212 to a V81, V121, or V201 Microscope. (305 and 508 Stemi includes adapter if purchased from Mitegen)
Catalog No. 50-004-95022 Supplier Mitegen Supplier No. ZVAXC212
The ZEISS Axiocam 212 is a smart 12-megapixel color microscope camera capable of delivering images at full 4K resolution and up to 30 frames per second. This camera can be used these MiTeGen Stereo Microscope Kits, Stemi 305, Stemi 508, V81, V121, or V201, when combined with the Axiocam Camera Adapter. In stand-alone mode, the camera automatically adjusts brightness and white balance and offers live image enhancement functions like sharpening, denoising, and HDR without the need for a PC. Alternatively, the CMOS camera can be connected to a computer via USB, Ethernet, or wirelessly via Wi-Fi dongle, making it an ideal solution for digital classroom applications and connected laboratories. SKU for adapter is ZV-Erc5-Adapter, and is necessary to mount the Axiocam 212 to a V81, V121, or V201 Microscope. (305 and 508 Stemi includes adapter if purchased from Mitegen)