Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axiom™ Apple Genotyping Array

Catalog No. 550573
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-057-3 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-057-3 Supplier LIFE TECHNOLOGIES Supplier No. 550573
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Array Genotyping
Number of Arrays 96 arrays
Format 96-array Plate
Species Apple
For Use With (Application) Microarray Analysis
Quantity 1 plate

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.