Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axiom™ Asia Precision Medicine Research Array Kit, 96-format

Catalog No. 905423
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-542-3 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-542-3 Supplier LIFE TECHNOLOGIES Supplier No. 905423
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Type Asia Precision Medicine Research Array Kit
Array Genotyping
Number of Arrays 96 arrays
Format 96-array Plate
Content And Storage • One 96-format array plate. Store at 2–8°C.
• One Axiom 2.0 Reagent Kit ((includes reagents and controls for 1 x 96-format array plate). Storage can vary for each reagent; refer to product insert.
• One Axiom GeneTitan Consumables Kit (includes GeneTitan instrument consumables to process 1 x 96 array plate). Store at room temperature.

For Use With (Application) Microarray Analysis
Product Line Axiom
Quantity 1 kit
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.