Learn More
Axiom™ Bovine Genotyping v3 Array (384HT format)
Specifications
Specifications
| Number of Arrays | 384 arrays |
| Format | 384 HT-array Plate |
| Species | Bovine |
| Content And Storage | One plate containing 384 arrays, Store in refrigerator (2-8°C) |
| For Use With (Application) | Microarray Analysis |
| Quantity | 1 plate |
| Shipping Condition | Room temperature |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.