Learn More
Axiom™ Bovine Genotyping v3 Array (mini-96 format)
Specifications
Specifications
| Format | Mini 96-array Plate |
| Content And Storage | One plate containing 96 arrays |
| For Use With (Application) | Microarray Analysis |
| Quantity | 1 plate |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.