Learn More
Axiom™ BreedWheat 35K Genotyping Array (384HT format)
Specifications
Specifications
| Array | Genotyping |
| Format | 384 HT-array Plate |
| Species | Wheat |
| Content And Storage | One plate containing 384 arrays, Store in refrigerator (2-8°C) |
| Quantity | 1 plate |
| Shipping Condition | Room temperature |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.