Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axiom™ Multispecies Shrimp and Tilapia Genotyping Array (Axiom™ TilShrv1), 384HT format

Catalog No. 551185
Encompass_Preferred
Change view
Click to view available options
Quantity:
384 arrays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-118-5 384 arrays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-118-5 Supplier LIFE TECHNOLOGIES Supplier No. 551185
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Array Genotyping
Number of Arrays 384 arrays
Format 384 HT-array Plate
Species Multi-species: Shrimp and Tilapia
Content And Storage Store in refrigerator (2°C to 8°C)
Quantity 384 arrays
Shipping Condition Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.