Learn More
Axiom™ Multispecies Shrimp and Tilapia Genotyping Array (Axiom™ TilShrv1), mini-96 format
Specifications
Specifications
| Array | Genotyping |
| Number of Arrays | 96 arrays |
| Format | Mini 96-array Plate |
| Species | Multi-species: Shrimp and Tilapia |
| Content And Storage | Store in refrigerator (2°C to 8°C) |
| Quantity | 96 arrays |
| Shipping Condition | Room temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.