Learn More
World Precision Instrument Axis Adjustment Tool
SDP
Catalog No. 502521339
Encompass
  • Micromanipulator adjustment tool to modify the anti-drift tension
  • Refer to the manual for instructions on use
  • For use with the KITE and M3301 micromanipulators
Catalog No. 50-252-1339 Supplier World Precision Instrument Supplier No. 502105
  • Micromanipulator adjustment tool to modify the anti-drift tension
  • Refer to the manual for instructions on use
  • For use with the KITE and M3301 micromanipulators