Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Axitinib-13C,d3 | 1261432-00-1 | 97.8% | 390.48 g·mol⁻1 | C2113CH15D3N4OS | 1 MG
SDP

Catalog No. 5000214278
Encompass_Preferred

Axitinib-13C,d3 is an isotopically labeled reference standard of axitinib, containing carbon-13 and three deuterium labels. It is intended for use as an internal standard, tracer, and reference material in analytical, metabolic, and pharmacokinetic studies, and is supplied as characterized, high-purity material with supporting documentation.

  • Isotopically labeled with 13C and d3 for tracer and internal standard use.
  • Purity 97.8% as reported by manufacturer.
  • Molecular weight 390.48 g·mol⁻1.
  • Provided as small-quantity reference material suitable for LC-MS workflows.
  • Supplied with batch-specific certificate of analysis for traceability.

Catalog No. 50-002-14278 Supplier Medchemexpress LLC Supplier No. HY10065S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Axitinib-13C,d3 is an isotopically labeled reference standard of axitinib, containing carbon-13 and three deuterium labels. It is intended for use as an internal standard, tracer, and reference material in analytical, metabolic, and pharmacokinetic studies, and is supplied as characterized, high-purity material with supporting documentation.

  • Isotopically labeled with 13C and d3 for tracer and internal standard use.
  • Purity 97.8% as reported by manufacturer.
  • Molecular weight 390.48 g·mol⁻1.
  • Provided as small-quantity reference material suitable for LC-MS workflows.
  • Supplied with batch-specific certificate of analysis for traceability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.