Learn More
Cayman Chemical Axitinib-13C-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1261432-00-1, 1 mg
SDP
Catalog No. 502934372
Encompass_Preferred
An internal standard for the quantification of axitinib by GC- or LC-MS
Catalog No. 50-293-4372 Supplier Cayman Chemical Supplier No. 266761 MG
An internal standard for the quantification of axitinib by GC- or LC-MS