Learn More
Cayman Chemical AxItInIb-13C-d3 500ug
SDP
Catalog No. 502934522
Encompass_Preferred
An internal standard for the quantification of axitinib by GC- or LC-MS
Catalog No. 50-293-4522 Supplier Cayman Chemical Supplier No. 26676500 UG
An internal standard for the quantification of axitinib by GC- or LC-MS