Learn More
Medchemexpress LLC HY-10065 200mg Medchemexpress, Axitinib CAS:319460-85-0 Purity:>98%
SDP
Catalog No. 502028632
Encompass_Preferred
Medchemexpress, HY-10065 200mg Axitinib CAS:319460-85-0 Axitinib is a multi-targeted tyrosine kinase inhibitor with IC50s of 0.1, 0.2, 0.1-0.3, 1.6 nM for VEGFR1, VEGFR2, VEGFR3 and PDGFRβ, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8632 Supplier Medchemexpress LLC Supplier No. HY10065200MG
Medchemexpress, HY-10065 200mg Axitinib CAS:319460-85-0 Axitinib is a multi-targeted tyrosine kinase inhibitor with IC50s of 0.1, 0.2, 0.1-0.3, 1.6 nM for VEGFR1, VEGFR2, VEGFR3 and PDGFRβ, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.