Learn More
Apexbio Technology LLC Axitinib (AG 013736) 319460-85-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061659
Encompass_Preferred
Axitinib (CAS 319460-85-0) also known as AG 013736 is an orally bioavailable selective inhibitor targeting vascular endothelial growth factor receptor (VEGFR) tyrosine kinases 1 2 and 3 It inhibits VEGFR phosphorylation with IC50 values of approximately 0 1 nM (VEGFR-1) 0 2 nM (VEGFR-2) and 0 1 0 3 nM (VEGFR-3) and demonstrates significantly higher selectivity relative to FGFR-1 Axitinib reduces phosphorylation of VEGF-mediated intracellular signaling proteins Akt eNOS and ERK1/2 and exhibits in vivo suppression of VEGFR-2 activation (EC50 0 49 nM) Its antiangiogenic action results in tumor growth delay in human xenograft models (e g HCT-116 SN12C) highlighting its utility in cancer research
Catalog No. 50-000-61659 Supplier Apexbio Technology LLC Supplier No. A83705.1
Axitinib (CAS 319460-85-0) also known as AG 013736 is an orally bioavailable selective inhibitor targeting vascular endothelial growth factor receptor (VEGFR) tyrosine kinases 1 2 and 3 It inhibits VEGFR phosphorylation with IC50 values of approximately 0 1 nM (VEGFR-1) 0 2 nM (VEGFR-2) and 0 1 0 3 nM (VEGFR-3) and demonstrates significantly higher selectivity relative to FGFR-1 Axitinib reduces phosphorylation of VEGF-mediated intracellular signaling proteins Akt eNOS and ERK1/2 and exhibits in vivo suppression of VEGFR-2 activation (EC50 0 49 nM) Its antiangiogenic action results in tumor growth delay in human xenograft models (e g HCT-116 SN12C) highlighting its utility in cancer research