Learn More
Apexbio Technology LLC Axitinib (AG 013736), 50mg. Cas: 319460-85-0 MFCD: MFCD09837898. VEGFR1/ c-Kit inhibitor
SDP
Supplier:  Apexbio Technology LLC A837050
Encompass_Preferred
Axitinib is a selective and oral inhibitor of vascular endothelial growth factor (VEGF) receptor tyrosine kinases 1, 2 and 3 with [1].Axitinib inhibited the phosphorylation of VEGFR-1, 2 and 3 with IC50 values of 1.2 nM, 0.2 nM and 0.1 to 0.3 nM in cells, respectively. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5131