Learn More
Selleck Chemical LLC Axitinib 50mg 319460-85-0 AG 013736
SDP
Catalog No. 506873
Encompass_Preferred
Axitinib (AG 013736) is a multi-target inhibitor of VEGFR1, VEGFR2, VEGFR3, PDGFR? and c-Kit with IC50 of 0.1 nM, 0.2 nM, 0.1-0.3 nM, 1.6 nM and 1.7 nM in Porcine aorta endothelial cells, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-687-3 Supplier Selleck Chemical LLC Supplier No. S100550MG
Axitinib (AG 013736) is a multi-target inhibitor of VEGFR1, VEGFR2, VEGFR3, PDGFR? and c-Kit with IC50 of 0.1 nM, 0.2 nM, 0.1-0.3 nM, 1.6 nM and 1.7 nM in Porcine aorta endothelial cells, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.