Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Axitinib-d3 | 1126623-89-9 | 98.1% | 389.49 g/mol | C22H15D3N4OS | 5 MG
SDP

Catalog No. 5000214275
Encompass_Preferred

Axitinib-d3 is a deuterium-labeled analog of axitinib supplied as an analytical standard for research and quantitative assays. It is intended as an internal standard or reference material for LC-MS and other analytical methods.

  • Deuterium-labeled internal standard for quantitative assays.
  • High chemical purity: 98.1%.
  • Chemical formula C22H15D3N4OS; molecular weight 389.49 g/mol.
  • CAS number 1126623-89-9 for unambiguous identification.
  • Suitable for LC-MS, stability studies, and method development.

Catalog No. 50-002-14275 Supplier Medchemexpress LLC Supplier No. HY10065S15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Axitinib-d3 is a deuterium-labeled analog of axitinib supplied as an analytical standard for research and quantitative assays. It is intended as an internal standard or reference material for LC-MS and other analytical methods.

  • Deuterium-labeled internal standard for quantitative assays.
  • High chemical purity: 98.1%.
  • Chemical formula C22H15D3N4OS; molecular weight 389.49 g/mol.
  • CAS number 1126623-89-9 for unambiguous identification.
  • Suitable for LC-MS, stability studies, and method development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.