Learn More
Medchemexpress LLC Axitinib-d3 | 1126623-89-9 | MFCD28138203 | 98.0% | 389.49 | C22H15D3N4OS | 1 MG
SDP
Catalog No. 5000212630
Encompass_Preferred
Axitinib-d3 is a deuterium-labeled analogue of axitinib supplied as a high-purity analytical standard. It is intended for use as a tracer or internal standard in quantitative methods such as NMR, GC-MS, and LC-MS, supporting accurate measurement in pharmacokinetic and bioanalytical studies.
  • High purity (98.0%) for reliable analytical performance.
  • Deuterium-labeled structure provides isotope differentiation from native compound.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS assays.
  • Useful in pharmacokinetic, ADME, and quantitative bioanalysis workflows.
  • Supplied as a small quantity vial (1 MG) for low-quantity applications.
Catalog No. 50-002-12630 Supplier Medchemexpress LLC Supplier No. HY10065S11MG
Axitinib-d3 is a deuterium-labeled analogue of axitinib supplied as a high-purity analytical standard. It is intended for use as a tracer or internal standard in quantitative methods such as NMR, GC-MS, and LC-MS, supporting accurate measurement in pharmacokinetic and bioanalytical studies.
  • High purity (98.0%) for reliable analytical performance.
  • Deuterium-labeled structure provides isotope differentiation from native compound.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS assays.
  • Useful in pharmacokinetic, ADME, and quantitative bioanalysis workflows.
  • Supplied as a small quantity vial (1 MG) for low-quantity applications.