Learn More
Medchemexpress LLC AXKO-0046 dihydrochloride | 99.7% | 448.47 | 1 ML
SDP
Catalog No. 5000363616
Encompass_Preferred
AXKO-0046 dihydrochloride, an indole derivative, is an uncompetitive and selective inhibitor of Lactate dehydrogenase B (LDHB). It demonstrates LDHB inhibitory activity with an EC50 value of 42 nM and is suitable for research in cancer metabolism.
  • Indole derivative
  • Uncompetitive Lactate dehydrogenase B (LDHB) selective inhibitor
  • Demonstrates LDHB inhibitory activity with an EC50 value of 42 nM
  • Can be utilized for research in cancer metabolism
Catalog No. 50-003-63616 Supplier Medchemexpress LLC Supplier No. HY147216A10MM/1ML
AXKO-0046 dihydrochloride, an indole derivative, is an uncompetitive and selective inhibitor of Lactate dehydrogenase B (LDHB). It demonstrates LDHB inhibitory activity with an EC50 value of 42 nM and is suitable for research in cancer metabolism.
  • Indole derivative
  • Uncompetitive Lactate dehydrogenase B (LDHB) selective inhibitor
  • Demonstrates LDHB inhibitory activity with an EC50 value of 42 nM
  • Can be utilized for research in cancer metabolism