Learn More
Medchemexpress LLC AXKO-0046 dihydrochloride | 99.71% | 448.47 | 25 MG
SDP
Catalog No. 5000362584
Encompass_Preferred
AXKO-0046 dihydrochloride is an indole derivative that acts as an uncompetitive and selective inhibitor of Lactate dehydrogenase B (LDHB). This compound is suitable for research related to cancer metabolism. It exhibits LDHB inhibitory activity with an EC50 value of 42 nM.
  • Indole derivative
  • Uncompetitive and selective LDHB inhibitor
  • EC50 value of 42 nM for LDHB inhibitory activity
  • Suitable for research in cancer metabolism
Catalog No. 50-003-62584 Supplier Medchemexpress LLC Supplier No. 25MGHY147216A
AXKO-0046 dihydrochloride is an indole derivative that acts as an uncompetitive and selective inhibitor of Lactate dehydrogenase B (LDHB). This compound is suitable for research related to cancer metabolism. It exhibits LDHB inhibitory activity with an EC50 value of 42 nM.
  • Indole derivative
  • Uncompetitive and selective LDHB inhibitor
  • EC50 value of 42 nM for LDHB inhibitory activity
  • Suitable for research in cancer metabolism