Learn More
ACROBiosystems ACROBiosystems Cynomolgus Axl Protein, His Tag
Supplier:  ACROBiosystems AXLC52H31MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus Axl, His Tag (AXL-C52H3) is expressed from human 293 cells (HEK293). It contains AA Pro 27 - Pro 293 (Accession # XP_005589384.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 30.7 kDa. The protein migrates as 42-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6667