Learn More
Medchemexpress LLC AXL Human HEK293 10ug
SDP
Catalog No. 5000178746
Encompass_Preferred
AXL protein is a receptor tyrosine kinase that mediates signals by binding to GAS6 and regulates cell survival proliferation migration and differentiation After ligand binding AXL dimerizes and autophosphorylates activating downstream molecules such as PI3 kinase subunits GRB2 PLCG1 etc AXL Protein Human (HEK293 His) is the recombinant human-derived AXL protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-78746 Supplier Medchemexpress LLC Supplier No. HYP7440710UG
AXL protein is a receptor tyrosine kinase that mediates signals by binding to GAS6 and regulates cell survival proliferation migration and differentiation After ligand binding AXL dimerizes and autophosphorylates activating downstream molecules such as PI3 kinase subunits GRB2 PLCG1 etc AXL Protein Human (HEK293 His) is the recombinant human-derived AXL protein expressed by HEK293 with C-His labeled tag