Learn More
Medchemexpress LLC AXL Mouse HEK293 100ug
SDP
Catalog No. 5000178873
Encompass_Preferred
The AXL protein has predicted roles in various cellular functions binds to phosphatidylinositol 3-kinase and phosphatidylserine and serves as a viral receptor It negatively regulates apoptosis and positively regulates protein kinase B signaling AXL Protein Mouse (HEK293 His-Fc) is the recombinant mouse-derived AXL protein expressed by HEK293 with C-hFc C-His labeled tag
Catalog No. 50-001-78873 Supplier Medchemexpress LLC Supplier No. HYP74406100UG
The AXL protein has predicted roles in various cellular functions binds to phosphatidylinositol 3-kinase and phosphatidylserine and serves as a viral receptor It negatively regulates apoptosis and positively regulates protein kinase B signaling AXL Protein Mouse (HEK293 His-Fc) is the recombinant mouse-derived AXL protein expressed by HEK293 with C-hFc C-His labeled tag