Learn More
Medchemexpress LLC AXL Mouse HEK293 50ug
SDP
Catalog No. 5000178538
Encompass_Preferred
The AXL protein has predicted roles in various cellular functions binds to phosphatidylinositol 3-kinase and phosphatidylserine and serves as a viral receptor It negatively regulates apoptosis and positively regulates protein kinase B signaling AXL Protein Mouse (HEK293 His-Fc) is the recombinant mouse-derived AXL protein expressed by HEK293 with C-hFc C-His labeled tag
Catalog No. 50-001-78538 Supplier Medchemexpress LLC Supplier No. HYP7440650UG
The AXL protein has predicted roles in various cellular functions binds to phosphatidylinositol 3-kinase and phosphatidylserine and serves as a viral receptor It negatively regulates apoptosis and positively regulates protein kinase B signaling AXL Protein Mouse (HEK293 His-Fc) is the recombinant mouse-derived AXL protein expressed by HEK293 with C-hFc C-His labeled tag