Learn More
Apexbio Technology LLC AXL1717 477-47-4 10mg
SDP
Catalog No. 502240074
Encompass_Preferred
AXL1717 (CAS 477-47-4) also known as picropodophyllin is a selective inhibitor of the insulin-like growth factor 1 receptor (IGF-1R) tyrosine kinase exhibiting an IC50 of 0 24 0 33 M IGF-1R a transmembrane receptor activated by IGF-1 is implicated in cellular proliferation and survival and its overexpression is associated with various malignancies AXL1717 demonstrates dose-dependent inhibition of cell viability induces G2-M cell cycle arrest and promotes apoptosis in multiple myeloma and colon cancer cell lines In murine xenograft models combination with HDAC inhibitor LBH589 prolongs survival AXL1717 has progressed to phase I/II trials in non-small cell lung cancer
This item is not returnable. View return policy
Catalog No. 50-224-0074 Supplier Apexbio Technology LLC Supplier No. A320910
AXL1717 (CAS 477-47-4) also known as picropodophyllin is a selective inhibitor of the insulin-like growth factor 1 receptor (IGF-1R) tyrosine kinase exhibiting an IC50 of 0 24 0 33 M IGF-1R a transmembrane receptor activated by IGF-1 is implicated in cellular proliferation and survival and its overexpression is associated with various malignancies AXL1717 demonstrates dose-dependent inhibition of cell viability induces G2-M cell cycle arrest and promotes apoptosis in multiple myeloma and colon cancer cell lines In murine xenograft models combination with HDAC inhibitor LBH589 prolongs survival AXL1717 has progressed to phase I/II trials in non-small cell lung cancer