Learn More
Enzo Life Sciences Axltide (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP2480001
Encompass_Preferred
This peptide, KKSRGDYMTMQIG, is derived from mouse insulin receptor 1 and is a substrate for Axl, DDR2, Mst1, and JAK2 kinases. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in Water. Long Term Storage: -20°C.
Catalog No. 50-201-1312