Learn More
Enzo Life Sciences Axltide (biotinylated) (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP2490001
Encompass_Preferred
This peptide, biotin-Ahx-KKSRGDYMTMQIG, is derived from mouse insulin receptor 1 and is a substrate for Axl, DDR2, Mst1, and JAK2 kinases. The biotin allows peptide to be used in kinase assays with streptavidin-bound membranes. Purity: ≥95%. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1313