Description
An SBS standard 96-well sitting-drop, high-throughput protein crystallization plate.
- Made of optically clear polystyrene to enable precise crystal visualization
- The buffer well has a novel D-shape geometry located on the right, with elevated round shape protein well located to its left
- Can be filled manually with pipetters or with automated liquid handling workstations
- The plate is sealed with commercially available clear adhesive seals, including Axygen™ UltraClear film
- Compatible with manual pipetters and automated robotic equipments
- Resistant to organic solvents
- Reduced polarization and static
- Plates are stackable
- Bar-coding option
- Round protein well allows easier loop access for crystal manipulation
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Color | Clear |
| Material | Polystyrene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Includes | 96 novel D-shape geometry 250μL buffer wells and an adjacent elevated round shape 4 to 3μL protein well |