Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 10μL Microvolume Filter Tips

Catalog No. 14222870 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-222-870 0.5 to 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-222-870 Supplier Axygen™ Supplier No. TXLF10

Axygen Universal Tips are designed to fit a wide variety of single and multi-channel pipettes, compatible with most popular brands of pipettes. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.

  • Technologically advanced filter material allows free passage of air while blocking aerosol contamination.
  • Filter membrane contains no leachable additives.
  • Axygen™ Maxymum Recovery™ surface improves accuracy and ensures maximum sample recovery.
  • RNase-/DNase-free
  • Nonpyrogenic
  • Human gDNA- and PCR-inhibition-free
  • TRUSTED_SUSTAINABILITY

    Specifications

    Volume (Metric) 0.5 to 10 μL
    Volume (Metric) Dispensing 0.5 to 10 μL
    Filtered Filtered
    Sterility Non-sterile
    Color Clear
    Certifications/Compliance USP 3127
    Clean Claims DNase, Rnase Free
    For Use With (Application) Allows recovery of costly samples with improved precision and accuracy
    Format Bulk
    Tip Style Extended Length, Microvolume
    Packaging 1,000 Tips/Pk., 10 Packs/Cs.
    Quantity 1000/Pk.
    Product Type Filtered Pipette Tips
    Includes 10 Racks x 96 Tips
    Green Features No Intentionally Added PFAS
    ACT Label No
    Disposable Single-use
    For Use With (Equipment) Gilson Pipetman P10, Gilson Pipetman P2
    Graduated No
    Material Polypropylene
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.