Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 10μL Microvolume Pipette Tips
GSA_VA

Catalog No. 14222743 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-222-743 0.5 to 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-222-743 Supplier Axygen™ Supplier No. T300RS

missing translation for 'validMainframeMsg'

Axygen Universal Tips are designed to fit a wide variety of single and multi-channel pipettes, compatible with most popular brands of pipettes. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.

  • Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free
  • Nonpyrogenic
  • Microvolume
  • Maxymum Recovery technology for ultra-low retention and high yield

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.5 to 10 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Model T-300-R-S
Certifications/Compliance USP 3127
Clean Claims DNase, Rnase Free, Nonpyrogenic
For Use With (Application) Allows recovery of costly samples with improved precision and accuracy
Format Rack
Tip Style Micro Point
Quantity 960/Pk., 4800/Cs.
Product Type Pipette Tips
For Use With (Equipment) Gilson Pipetman P10, Gilson Pipetman P2
Product Line Maxymum Recovery
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.